accessibility menu, dialog, popup

UserName

CAS RN 563-41-7

IUPAC Name: hydrogen aminourea chloride
Synonyms: hydrogen semicarbazide chloride
Molecular Weight (g/mol): 111.53
Molecular Formula: CH6ClN3O
InChi Key: XHQYBDSXTDXSHY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC(N)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)