Learn More
Semicarbazide hydrochloride, 99%
Urease substrate and MAO inhibitor | CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol
Supplier: Thermo Scientific Chemicals A116680E
Description
Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSemicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.
Solubility
Soluble in water (>100g/L).
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
| Semicarbazide hydrochloride | |
| ∼176°C (decomposition) | |
| Odorless | |
| H2NCONHNH·HCl | |
| 2500 g | |
| 3593642 | |
| semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
| XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
| aminourea;hydrochloride | |
| 11236 | |
| 111.53 | |
| Crystalline |
| 563-41-7 | |
| 1.5 | |
| CH6ClN3O | |
| MFCD00013009 | |
| UN2811 | |
| 14,8444 | |
| Soluble in water (>100g/L). | |
| [H+].[Cl-].NNC(N)=O | |
| 111.53 | |
| CHEBI:82532 | |
| 99% |
Chemical Identifiers
| 563-41-7 | |
| 111.53 | |
| XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
| 11236 | |
| aminourea;hydrochloride |
| CH6ClN3O | |
| MFCD00013009 | |
| semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
| CHEBI:82532 | |
| [H+].[Cl-].NNC(N)=O |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P264b-P270-P280-P280i-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
H301-H315-H319-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 209-247-0
RTECSNumber : VT3500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only