missing translation for 'onlineSavingsMsg'
Learn More

Semicarbazide hydrochloride, 99%

Catalog No. AAA1166822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1166822 100 g
AAA1166836 500 g
AAA116680E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1166822 Supplier Thermo Scientific Chemicals Supplier No. A1166822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Urease substrate and MAO inhibitor | CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol

Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.

Solubility
Soluble in water (>100g/L).

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 563-41-7
Molecular Formula CH6ClN3O
Molecular Weight (g/mol) 111.53
MDL Number MFCD00013009
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
PubChem CID 11236
ChEBI CHEBI:82532
IUPAC Name aminourea;hydrochloride
SMILES [H+].[Cl-].NNC(N)=O

Specifications

Melting Point ∼176°C (decomposition)
Color White
pH 1.5
Odor Odorless
Linear Formula H2NCONHNH·HCl
Quantity 100 g
UN Number UN2811
Beilstein 3593642
Merck Index 14,8444
Solubility Information Soluble in water (>100g/L).
Formula Weight 111.53
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Semicarbazide hydrochloride
Show More Show Less
Hazard Category H301-H315-H319-H351
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P270-P280-P280i-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-247-0
RTECSNumber VT3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only