missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semen Diluting Fluid, Bicarbonate-Formalin Diluting Fluid for Sperm Count, Ricca Chemical
$114.53 - $114.53
Specifications
| Chemical Name or Material | Semen Diluting Fluid |
|---|---|
| Color | Colorless |
| Name Note | Bicarbonate-Formalin Diluting Fluid for Sperm Count |
| CAS | 7732-18-5,144-55-8,50-00-0 |
| CAS Min % | 92.0,4.62,0.99 |
Specifications
| Semen Diluting Fluid | |
| Bicarbonate-Formalin Diluting Fluid for Sperm Count | |
| 92.0,4.62,0.99 | |
| Laboratory | |
| Liquid |
| Colorless | |
| 7732-18-5,144-55-8,50-00-0 | |
| 96.0,4.81,1.03 | |
| Natural Poly Bottle |