missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581009
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SEMA6A.
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.Chemical Identifiers
| SEMA6A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| SEMA6A | |
| 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
| SEMA6A | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 1 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Q9H2E6 | |
| 57556 | |
| Recombinant Human SEMA6A Protein (Met1-Thr649). | |
| Antibody |
Specifications
| SEMA6A | |
| Polyclonal | |
| Unconjugated | |
| SEMA6A | |
| 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9H2E6 | |
| SEMA6A | |
| Recombinant Human SEMA6A Protein (Met1-Thr649). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |