missing translation for 'onlineSavingsMsg'
Learn More

Selleger's Stain, Reagents

Selleger's Stain, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$235.04 - $881.25

Chemical Identifiers

CAS 13477-34-4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01333896
View Documents Promotion Details
N/A 30 mL
Each for $235.04
Only null left
 
01333895
View Documents Promotion Details
N/A 500 mL
Each for $881.25
Only null left
 
01333894
View Documents Promotion Details
N/A 50 mL
Each for $293.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13477-34-4

Specifications

13477-34-4
Liquid
Glass Bottle
MWGCGFYACDTFSB-UHFFFAOYSA-N
calcium bis(nitric acid) tetrahydrate
Completely miscible
Brown
CaH8N2O10
Glass Bottle
O.[Ca++].[O-][N+]([O-])=O
236.15
Videos
SDS
Documents

Documents