missing translation for 'onlineSavingsMsg'
Learn More

Selenous acid, 97%

Catalog No. p-4408644
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1257406 5 g
AA1257418 50 g
AA1257430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1257406 Supplier Thermo Scientific Chemicals Supplier No. 01257406

For Use As An Oxidizing Agent, As a Reagent For Alkaloids | CAS: 7783-00-8 | H2O3Se | 128.98 g/mol

As an oxidizing agent, as a reagent for alkaloidsSelenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-00-8
Molecular Formula H2O3Se
Molecular Weight (g/mol) 128.98
MDL Number MFCD00011331
InChI Key MCAHWIHFGHIESP-UHFFFAOYSA-N
Synonym selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution
PubChem CID 1091
ChEBI CHEBI:26642
IUPAC Name selenous acid
SMILES O[Se](O)=O

Specifications

Color Colorless
Melting Point 70°C (decomposition)
Density 3.004g/cm3 at 20°C
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 97%
Quantity 5 g
Physical Form -4 Mesh Crystalline
Solubility Information Freely soluble in water,alcohol. Insoluble in ammoniaSoluble in water and ethanol.
Formula Weight 128.97
Chemical Name or Material Selenous acid
Show More Show Less
Hazard Category H301+H331-H373
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 231-974-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only