CAS RN 7783-00-8
Selenous acid, Puratronic™, 99.999% (metals basis)
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N Synonym: selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
Selenious acid, 99.999%, (trace metal basis)
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N Synonym: selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
Selenous acid, 97%
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N Synonym: selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
Selenious acid, 98%
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N Synonym: selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
VeriSpec™ Se4+ Standard for ICP 100 ppm in 2% HNO3, Ricca Chemical
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
VeriSpec™ Selenium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
VeriSpec™ Selenium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
VeriSpec™ Selenium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O
VeriSpec™ Selenium (Se) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 7783-00-8 Molecular Formula: H2O3Se Molecular Weight (g/mol): 128.98 MDL Number: MFCD00011331 InChI Key: MCAHWIHFGHIESP-UHFFFAOYSA-N PubChem CID: 1091 ChEBI: CHEBI:26642 IUPAC Name: selenous acid SMILES: O[Se](O)=O