missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Selenoprotein W Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149818
Description
Selenoprotein W Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a selenoprotein containing a selenocysteine (Sec) residue, which is encoded by the UGA codon that normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, the Sec insertion sequence (SECIS) element that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein is highly expressed in skeletal muscle, heart and brain. It belongs to the SelWTH family, which possesses a thioredoxin-like fold and a conserved CxxU (C is cysteine, U is Sec) motif, suggesting a redox function for this gene. Studies in mouse show that this selenoprotein is involved in muscle growth and differentiation, and in the protection of neurons from oxidative stress during neuronal development. A retroprocessed pseudogene for this locus has been identified on chromosome 1.Specifications
| Selenoprotein W | |
| Polyclonal | |
| Unconjugated | |
| SELENOW | |
| Selenoprotein W; selenoprotein W, 1; selenoprotein W, muscle 1; Selenow; SelW; Sepw1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6415 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P63302 | |
| SELENOW | |
| Recombinant Selenoprotein W1 (SEPW1) (Met1-Gly87). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |