missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Seleno-L-methionine

CAS: 3211-76-5 | C5H11NO2Se | 196.12 g/mol

Supplier:  Thermo Scientific Chemicals J6690203

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6690203


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3211-76-5
196.12
RJFAYQIBOAGBLC-BYPYZUCNSA-N
105024
(2S)-2-amino-4-methylselanylbutanoic acid
C5H11NO2Se
MFCD00037210
l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid
CHEBI:62621
C[Se]CC[C@H](N)C(O)=O

Specifications

265°C
3211-76-5
C5H11NO2Se
MFCD00037210
l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid
RJFAYQIBOAGBLC-BYPYZUCNSA-N
(2S)-2-amino-4-methylselanylbutanoic acid
1 g
CHEBI:62621
Powder
Seleno-L-methionine
White
CH3SeCH2CH2CH(NH2)CO2H
UN3283
Soluble in water
C[Se]CC[C@H](N)C(O)=O
196.12
105024
196.11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

RTECSNumber : EK7713840

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions