missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selenium AA Standard, 1000 ppm Se in H2O, Ricca Chemical
$141.63 - $337.94
Chemical Identifiers
| CAS | 7446-08-4 |
|---|---|
| Molecular Formula | O2Se |
| Molecular Weight (g/mol) | 110.97 |
| MDL Number | MFCD00003562 |
| InChI Key | JPJALAQPGMAKDF-UHFFFAOYSA-N |
| PubChem CID | 24007 |
| IUPAC Name | (oxo-λ4-selanylidene)oxidane |
| SMILES | O=[Se]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ASE1KW-100
|
Ricca Chemical Company
ASE1KW100 |
100 mL | Natural LDPE Bottle |
Each for $141.63
|
|
||||
|
ASE1KW-500
|
Ricca Chemical Company
ASE1KW500 |
500 mL | Natural Poly Bottle |
Each for $337.94
|
|
||||
Chemical Identifiers
| 7446-08-4 | |
| 110.97 | |
| JPJALAQPGMAKDF-UHFFFAOYSA-N | |
| (oxo-λ4-selanylidene)oxidane |
| O2Se | |
| MFCD00003562 | |
| 24007 | |
| O=[Se]=O |
Specifications
| Natural LDPE Bottle | |
| 98.0,0.14 | |
| 100.0,0.14 | |
| Liquid | |
| MFCD00003562 | |
| 100°C | |
| O=[Se]=O | |
| 110.97 | |
| 24007 |
| 7446-08-4,7732-18-5 | |
| 1,000ppm Se | |
| 1 mL = 1 mg Se | |
| O2Se | |
| Colorless | |
| JPJALAQPGMAKDF-UHFFFAOYSA-N | |
| (oxo-λ4-selanylidene)oxidane | |
| 100 mL | |
| Selenium AA Standard |