missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Sodium tetrakis(4-fluorophenyl)borate dihydrate, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111013514
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013514 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013514 Supplier Merck Emd Millipore Supplier No. 72014500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium tetrakis(4-fluorophenyl)borate dihydrate is sodium salt derivative of tetraphenylborate (TPB).

Chemical Identifiers

MDL Number MFCD00149598
Synonym Cesibor; Tetrakis(4-fluorophenyl)boron sodium

Specifications

CAS 207683-22-5
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula (FC6H4)4 BNa·2H2 O
Quantity 500 mg
Formula Weight 450.21
Percent Purity ≥97.0% (NT); ≥97.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH