missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Sodium ionophore III, Function Tested, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Sodium ionophore III is generally used for preparing ion-selective electrode.
$616.02 - $2519.34
Chemical Identifiers
| MDL Number | MFCD00077941 |
|---|---|
| Synonym | ETH 2120; N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide |
Chemical Identifiers
| MFCD00077941 | |
| ETH 2120; N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide |
Specifications
| 81686-22-8 | |
| C34H52N2O4 | |
| 50 mg | |
| 552.79 | |
| Selectophore |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00077941 | |
| ETH 2120; N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide | |
| ≥98.0% (HPLC) | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH