missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Sodium ionophore III, Function Tested, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015613
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015613 50 mg
111015612 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015613 Supplier Merck Emd Millipore Supplier No. 7173450MG

Sodium ionophore III is generally used for preparing ion-selective electrode.

Chemical Identifiers

MDL Number MFCD00077941
Synonym ETH 2120; N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide

Specifications

CAS 81686-22-8
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C34H52N2O4
Quantity 50 mg
Formula Weight 552.79
Percent Purity ≥98.0% (HPLC)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH