missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Potassium tetrakis(4-chlorophenyl)borate, ≥98.0%, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$100.63 - $1491.53
Chemical Identifiers
| MDL Number | MFCD00043105 |
|---|---|
| Synonym | Tetrakis(4-chlorophenyl)boron potassium |
Chemical Identifiers
| MFCD00043105 | |
| Tetrakis(4-chlorophenyl)boron potassium |
Specifications
| 14680-77-4 | |
| (ClC6H4)4 BK | |
| 100 mg | |
| 496.11 | |
| Selectophore |
| Glass Bottle | |
| MFCD00043105 | |
| Tetrakis(4-chlorophenyl)boron potassium | |
| ≥98.0% (AT); ≥98.0% | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH