accessibility menu, dialog, popup

UserName

CAS RN 14680-77-4

IUPAC Name: boron(3+) potassium tetrakis(3-chlorobenzen-1-ide)
Synonyms: boron(3+) potassium tetrakis(3-chlorobenzen-1-ide)
Molecular Weight (g/mol): 496.1
Molecular Formula: C24H16BCl4K
InChi Key: VHPYLDPFVZBNQT-UHFFFAOYSA-N
SMILES: [B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)