missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥95.0% (based on K, ICP), MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$739.12 - $4126.63
Chemical Identifiers
| MDL Number | MFCD00077934 |
|---|---|
| Synonym | Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron potassium |
Chemical Identifiers
| MFCD00077934 | |
| Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron potassium |
Specifications
| 126°C to 129°C | |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00077934 | |
| Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron potassium | |
| ≥95.0% (based on K, ICP) | |
| Selectophore |
| 105560-52-9 | |
| C32H12 BF24 K | |
| 50 mg | |
| 902.31 | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH