missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥95.0% (based on K, ICP), MilliporeSigma™ Supelco™

Catalog No. 111012382
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012382 50 mg
111012381 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012382 Supplier Merck Emd Millipore Supplier No. 6058850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00077934
Synonym Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron potassium

Specifications

CAS 105560-52-9
Melting Point 126°C to 129°C
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C32H12 BF24 K
Quantity 50 mg
Molecular Weight (g/mol) 902.31
Formula Weight 902.31
Percent Purity ≥95.0% (based on K, ICP)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH