missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Potassium ionophore III, Function Tested, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$486.51 - $1,614.71
Chemical Identifiers
| MDL Number | MFCD00145150 |
|---|---|
| Synonym | 2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5 ′-nitro(benzo-15-crown-5)-4 ′-yl]carbamate]; BME 44 |
Chemical Identifiers
| MFCD00145150 | |
| 2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5 ′-nitro(benzo-15-crown-5)-4 ′-yl]carbamate]; BME 44 |
Specifications
| 99348-39-7 | |
| C46H70N4O18 | |
| 25 mg | |
| 967.06 | |
| Selectophore |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00145150 | |
| 2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5 ′-nitro(benzo-15-crown-5)-4 ′-yl]carbamate]; BME 44 | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH