missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Potassium ionophore III, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111016019
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016019 25 mg
111016018 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016019 Supplier Merck Emd Millipore Supplier No. 6039725MG

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00145150
Synonym 2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5 ′-nitro(benzo-15-crown-5)-4 ′-yl]carbamate]; BME 44

Specifications

CAS 99348-39-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C46H70N4O18
Quantity 25 mg
Formula Weight 967.06
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH