missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Poly(vinyl chloride), high molecular weight, MilliporeSigma™ Supelco™

$177.75 - $622.56

Chemical Identifiers

MDL Number MFCD00084459
Synonym PVC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015769
View Documents
Merck Emd Millipore
8139210G
10 g N/A
Only null left
 
111015770
View Documents
Merck Emd Millipore
8139250G
50 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00084459
PVC

Specifications

9002-86-2
(CH2 CHCl)n
10 g
Selectophore
Selectophore
1.4 g/mL (at 25°C)
MFCD00084459
PVC
Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KV0350000

SDS
missing translation for 'documents'

missing translation for 'documents'

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH