missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Poly(vinyl chloride), high molecular weight, MilliporeSigma™ Supelco™

Catalog No. 111015770
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015770 50 g
111015769 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015770 Supplier Merck Emd Millipore Supplier No. 8139250G
Only null left

Chemical Identifiers

MDL Number MFCD00084459
Synonym PVC

Specifications

Density 1.4 g/mL (at 25°C)
CAS 9002-86-2
Linear Formula (CH2 CHCl)n
Quantity 50 g
Grade Selectophore
Physical Form Powder
Product Line Selectophore
RTECSNumber KV0350000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH