missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Nitrate Ionophore VI, Function Tested, MilliporeSigma™ Supelco™
New ionophore for online monitoring of nitrate obtains superior characteristics over all existing nitrate ionophores.
$255.02 - $6425.93
Chemical Identifiers
| MDL Number | MFCD16652520 |
|---|---|
| Synonym | 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane |
Chemical Identifiers
| MFCD16652520 | |
| 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane |
Specifications
| 1196157-85-3 | |
| C28H60N4O4 | |
| 10 mg | |
| 516.80 | |
| Selectophore |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD16652520 | |
| 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH