Learn More
Selectophore™ Nitrate Ionophore VI, Function Tested, MilliporeSigma™ Supelco™
New ionophore for online monitoring of nitrate obtains superior characteristics over all existing nitrate ionophores.
Supplier: Merck Emd Millipore 07295500MG
Specifications
| 1196157-85-3 | |
| C28H60N4O4 | |
| 500 mg | |
| 516.80 | |
| Selectophore |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD16652520 | |
| 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane | |
| Selectophore |
Chemical Identifiers
| MFCD16652520 | |
| 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH