missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Magnesium Ionophore VI, Function Tested, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Lipophilic magnesium-selective ionophore (log PTLC=8.5) with sufficient selectivity for Mg2+ against the main interfering cations for serum measurments
$2242.55 - $13999.06
Chemical Identifiers
| CAS | 151058-38-7 |
|---|---|
| Molecular Formula | C63H96N6O6 |
| Molecular Weight (g/mol) | 1033.50 |
| MDL Number | MFCD01867417 |
| InChI Key | ODMHCQNFQKBJOO-UHFFFAOYSA-N |
| Synonym | 1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene; ETH 5506 |
| IUPAC Name | N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide |
| SMILES | CN(C(=O)CC(=O)NCCCCCC1=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=C1)C12CC3CC(CC(C3)C1)C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
111013202
|
Merck Emd Millipore
6311210MG |
10 mg | Bottomless Glass Bottle with Fused Cone |
Each for $2,242.55
|
|
||||
|
111013201
|
Merck Emd Millipore
63112100MG |
100 mg | Bottomless Glass Bottle with Fused Cone |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 151058-38-7 | |
| 1033.50 | |
| ODMHCQNFQKBJOO-UHFFFAOYSA-N | |
| N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide |
| C63H96N6O6 | |
| MFCD01867417 | |
| 1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene; ETH 5506 | |
| CN(C(=O)CC(=O)NCCCCCC1=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=C1)C12CC3CC(CC(C3)C1)C2 |
Specifications
| 151058-38-7 | |
| C63H96N6O6 | |
| MFCD01867417 | |
| 1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene; ETH 5506 | |
| CN(C(=O)CC(=O)NCCCCCC1=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=C1)C12CC3CC(CC(C3)C1)C2 | |
| 1033.50 | |
| ≥99.0% (HPLC) | |
| Selectophore |
| Bottomless Glass Bottle with Fused Cone | |
| C63H96N6O6 | |
| 10 mg | |
| ODMHCQNFQKBJOO-UHFFFAOYSA-N | |
| N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide | |
| 1033.47 | |
| Selectophore™ |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH