accessibility menu, dialog, popup

UserName

CAS RN 151058-38-7

IUPAC Name: N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide
Synonyms: N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide
Molecular Weight (g/mol): 1033.5
Molecular Formula: C63H96N6O6
InChi Key: ODMHCQNFQKBJOO-UHFFFAOYSA-N
SMILES: CN(C(=O)CC(=O)NCCCCCC1=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=C1)C12CC3CC(CC(C3)C1)C2
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (1)
  • (1)
Product Line 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)