missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Lithium Ionophore VI, Function Tested, 99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012402
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111012402 100 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012402 Supplier Merck Emd Millipore Supplier No. 62567100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106868-21-7
Molecular Formula C24H32O4
Molecular Weight (g/mol) 384.52
MDL Number MFCD00037498
InChI Key KDMUFHBMXZLDLI-UHFFFAOYSA-N
Synonym 6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4
IUPAC Name 6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane
SMILES C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C24H32O4
Quantity 100 mg
Formula Weight 384.51
Percent Purity ≥99.0% (HPLC)
Grade Selectophore
Physical Form Solid
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH