accessibility menu, dialog, popup

UserName

OOOO

CAS RN 106868-21-7

IUPAC Name: 6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane
Synonyms: 6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane
Molecular Weight (g/mol): 384.52
Molecular Formula: C24H32O4
InChi Key: KDMUFHBMXZLDLI-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1
Grade 
  • (1)
Percent Purity 
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)