missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Diphenyl ether, 99.9%, MilliporeSigma™ Supelco™

Catalog No. 111020174
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020174 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020174 Supplier Merck Emd Millipore Supplier No. 6733425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diphenyl ether (diphenyl oxide) belongs to the family of aromatic homomonocyclic compounds. It is colorless, exists as crystalline solid or liquid (above 82F) and has a geranium-like odor. Diphenyl ether is present in alcoholic beverages. It is used as a flavouring ingredient and is found in vanilla, green tea, muscat grapes, potato chips, grilled beef, buckwheat, and lemon balm. It is a starting material in the production of phenoxathiin via the Ferrario reaction.. In similar reactions, diphenyl ether is a significant side product in the high-pressure hydrolysis of chlorobenzene during production of phenol. Also several polybrominated diphenyl ethers (PBDEs) are useful flame retardants.

Chemical Identifiers

CAS 101-84-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00003034
InChI Key USIUVYZYUHIAEV-UHFFFAOYSA-N
Synonym Diphenyl oxide; Phenyl ether
IUPAC Name phenoxybenzene
SMILES O(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 25°C to 27°C; 27°C to 29°C
Density 1.073 g/mL (at 25°C)
Boiling Point 259°C
Linear Formula (C6H5)2 O
Refractive Index n20/D 1.579
Quantity 25 mL
Vapor Density >5.86 (25°C, vs. air)
Formula Weight 170.21
Vapor Pressure <1 mmHg (20°C)
Percent Purity ≥99.9%
Grade Selectophore
Product Line Selectophore
Show More Show Less
RTECSNumber KN8970000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH