missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Diisodecyl phthalate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019876
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019876 10 mL
111018377 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019876 Supplier Merck Emd Millipore Supplier No. 8013510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diisodecyl phthalate (DIDP) belongs to phthalate group, and is a dialkyl or aryl/alkyl diesters of phthalic acid. It is mostly used as a plasticizer in manufacturing polymers and other consumer products.

Specifications

CAS 26761-40-0
Density 0.965 g/mL (at 20°C)
Flash Point 275°C
Linear Formula C28H46O4
Quantity 10 mL
Formula Weight 446.66
Percent Purity ≥99.0% (sum of isomers, GC); ≥99.0%
Grade Selectophore
Product Line Selectophore
RTECSNumber TI1270000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH