missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Calcium Ionophore IV - Cocktail for membranes, MilliporeSigma™ Supelco™

Catalog No. 111016563
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016563 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016563 Supplier Merck Emd Millipore Supplier No. 703561MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

CAS 109-99-9

Specifications

Flash Point -18°C
Quantity 1 mL
Grade Selectophore
Physical Form Liquid
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH