missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Bis(2-ethylhexyl) adipate, ≥99.0%, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
Supplier: Merck Emd Millipore 0213825ML
Specifications
| <-70°C | |
| 0.925 g/mL (at 20°C) | |
| 196°C - Closed Cup | |
| MFCD00009496 | |
| Adipic acid di(2-ethylhexyl) ester; DOA | |
| ≥99.0% (GC); ≥99.0% | |
| Selectophore |
| 103-23-1 | |
| 175°C (2 mmHg) | |
| [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2 | |
| 25 mL | |
| 370.57 | |
| Selectophore |
Chemical Identifiers
| MFCD00009496 | |
| Adipic acid di(2-ethylhexyl) ester; DOA |
Safety and Handling
RTECSNumber : AU9700000
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH