missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Bis(2-ethylhexyl) adipate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187988
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012335 1 mL
111012336 5 mL
111012334 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111012335 Supplier Merck Emd Millipore Supplier No. 021381ML
Only null left

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00009496
Synonym Adipic acid di(2-ethylhexyl) ester; DOA

Specifications

Melting Point <-70°C
CAS 103-23-1
Density 0.925 g/mL (at 20°C)
Boiling Point 175°C (2 mmHg)
Flash Point 196°C - Closed Cup
Linear Formula [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 1 mL
Formula Weight 370.57
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Product Line Selectophore
Show More Show Less
RTECSNumber AU9700000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH