missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ 1-Octadecanol, ≥99.5%, MilliporeSigma™ Supelco™

Visit our Sensor Applications portal to learn more.

$257.88 - $257.88

Chemical Identifiers

MDL Number MFCD00002823
Synonym Octadecyl alcohol; Stearyl alcohol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012519
View Documents
Merck Emd Millipore
747235G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00002823
Octadecyl alcohol; Stearyl alcohol

Specifications

56°C to 59°C
0.812 g/mL (at 25°C)
842°F
MFCD00002823
Octadecyl alcohol; Stearyl alcohol
9.3 (vs. air)
<0.01 mmHg (38°C)
Selectophore
112-92-5
195°C
CH3(CH2)17 OH
5 g
THF: 1 g/10 mL, clear, colorless
270.49
≥99.5% (GC); ≥99.5%
Selectophore
Videos
Safety and Handling

Safety and Handling

RTECSNumber : RG2010000

SDS
missing translation for 'documents'

missing translation for 'documents'

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH