missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ 1-Octadecanol, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111012519
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012519 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012519 Supplier Merck Emd Millipore Supplier No. 747235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00002823
Synonym Octadecyl alcohol; Stearyl alcohol

Specifications

Melting Point 56°C to 59°C
Density 0.812 g/mL (at 25°C)
CAS 112-92-5
Flash Point 195°C
Ignition Point 842°F
Linear Formula CH3(CH2)17 OH
Quantity 5 g
Solubility Information THF: 1 g/10 mL, clear, colorless
Vapor Density 9.3 (vs. air)
Formula Weight 270.49
Vapor Pressure <0.01 mmHg (38°C)
Percent Purity ≥99.5% (GC); ≥99.5%
Grade Selectophore
Product Line Selectophore
Show More Show Less
RTECSNumber RG2010000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH