Learn More
Polymedco™ Sedimat™ 15 Plus Sedrate System
Fast, in-office results for one to eight specimens simultaneously.
$278.39 - $10,885.50
Specifications
| For Use With (Application) | Fast, in-office results for one to eight specimens simultaneously |
|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13641587B
|
Polymedco™
HT100 |
Thermal Printer Paper |
Each for $278.39
|
|
|||||
21000118
|
Polymedco™
SJ 1215 PLUS |
Sedimat 15 Plus Sedrate System |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Automated sedrate system provides accurate ESR results in only 15 minutes.
- Random access capability
- Faster throughput; processes 32 patient results per hour
- Eight-position reader
- Small footprint suitable for tight benchtops
- Bar code reader available
- LIS compatible for data transfer
- Printer available (21-000-123)
Specifications
| Fast, in-office results for one to eight specimens simultaneously |