Learn More
Polymedco™ Sedimat™ 15 Plus Sedrate System
Catalog No. 21000118
Encompass
Change view
Click to view available options
Type:
Sedimat 15 Plus Sedrate System
Thermal Printer Paper
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21000118 Sedimat 15 Plus Sedrate System
13641587B Thermal Printer Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21000118 Supplier Polymedco™ Supplier No. SJ 1215 PLUS
Fast, in-office results for one to eight specimens simultaneously.
Automated sedrate system provides accurate ESR results in only 15 minutes.
  • Random access capability
  • Faster throughput; processes 32 patient results per hour
  • Eight-position reader
  • Small footprint suitable for tight benchtops
  • Bar code reader available
  • LIS compatible for data transfer
  • Printer available (21-000-123)
Specifications
Type Sedimat 15 Plus Sedrate System
For Use With (Application) Fast, in-office results for one to eight specimens simultaneously
Sample Size 0.8 mL
Throughput 32 Samples/Hour