missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Securin Monoclonal Antibody (K49008_2B9), Invitrogen™

Catalog No. MA556155
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556155 200 μL
MA555775 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556155 Supplier Invitrogen™ Supplier No. MA556155
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MATLIYVDKE NGEPGTRVVA KDGLKLGSGP SIKALDGRSQ VSTPRFGKTF DAPPALPKAT RKALGTVNRA TEKSVKTKGP LKQKQPSFSA KKMTEKTVKA KSSVPASDDA YPEIEKFFPF NPLDFESFDL PEEHQIAHLP LSGVPLMILD EERELEKLFQ LGPPSPVKMP SPPWESNLLQ SPSSILSTLD VELPPVCCDI DI

The encoded protein is a homolog of yeast securin proteins, which prevent separins from promoting sister chromatid separation. It is an anaphase-promoting complex (APC) substrate that associates with a separin until activation of the APC. The gene product has transforming activity in vitro and tumorigenic activity in vivo, and the gene is highly expressed in various tumors. The gene product contains 2 PXXP motifs, which are required for its transforming and tumorigenic activities, as well as for its stimulation of basic fibroblast growth factor expression. It also contains a destruction box (D box) that is required for its degradation by the APC. The acidic C-terminal region of the encoded protein can act as a transactivation domain. The gene product is mainly a cytosolic protein, although it partially localizes in the nucleus.

Specifications

Antigen Securin
Applications Western Blot
Classification Monoclonal
Clone K49008_2B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Pttg1
Gene Accession No. O95997
Gene Alias AW555095; C87862; EAP1; Esp1-associated protein; ESP1-associated protein 1; HPTTG; MGC126883; MGC138276; Pituitary tumor transforming; pituitary tumor-transforming 1; pituitary tumor-transforming gene 1; pituitary tumor-transforming gene 1 protein; PTTG; PTTG1; Pttg3; Securin; tumor-transforming protein 1; TUTR1
Gene Symbols Pttg1
Host Species Mouse
Immunogen Recombinant human PTTG1 (immunogen range is 1-202).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less