Learn More
Restek™ Secure-Fit Fitting, Single Connector
Secure-Fit connectors ensure a consistent, leak-free seal and eliminate excess dead volume.
Supplier: Restek™ RK25182
Description
An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.- Material: Stainless-steel
Specifications
| Connector | |
| 0.005 in. | |
| 10 cm | |
| 10 cm |