Learn More
Restek™ Secure-Fit Fitting, Single Connector
Secure-Fit connectors ensure a consistent, leak-free seal and eliminate excess dead volume.
$282.19 - $282.19
Description
An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.- Material: Stainless-steel
Specifications
| Connector |