missing translation for 'onlineSavingsMsg'
Learn More

Secondary Drinking Water Metals For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC EP4

Catalog No. EP-4


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Zn500μg/mL
Cu100μg/mL
Fe30μg/mL
Mn5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Secondary Drinking Water Metals
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP
Certified Reference Material
125 mL
Components in 2% HNO3:
Cu    •100μg/mL
Fe    •30μg/mL
Mn    •5μg/mL
Zn    •500μg/mL
Triple-leached LDPE bottle
Liquid
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
2% HNO3
Environmental
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only