missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Methyl Ether 98.0+%, TCI America™
Supplier: TCI America B198525ML
Specifications
| sec-Butyl Methyl Ether | |
| Colorless | |
| C5H12O | |
| 25 mL | |
| sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a | |
| CCC(C)OC | |
| 88.15 | |
| 88.15 | |
| Liquid |
| 6795-87-5 | |
| 59°C | |
| MFCD00190205 | |
| 3271 | |
| FVNIMHIOIXPIQT-UHFFFAOYNA-N | |
| 2-methoxybutane | |
| 23238 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6795-87-5 | |
| 88.15 | |
| FVNIMHIOIXPIQT-UHFFFAOYNA-N | |
| 23238 | |
| CCC(C)OC |
| C5H12O | |
| MFCD00190205 | |
| sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a | |
| 2-methoxybutane |
Safety and Handling
RTECSNumber : KN5249500
TSCA : No