missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Methyl Ether 98.0+%, TCI America™

Catalog No. p-7119401
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
B19855ML 5 mL
B198525ML 25 mL
2 options

Catalog No. B19855ML

Supplier: TCI America B19855ML

Only null left

Chemical Identifiers

CAS 6795-87-5
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00190205
InChI Key FVNIMHIOIXPIQT-UHFFFAOYNA-N
Synonym sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
PubChem CID 23238
IUPAC Name 2-methoxybutane
SMILES CCC(C)OC

Specifications

Color Colorless
Boiling Point 59°C
Quantity 5 mL
UN Number 3271
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Methyl Ether
RTECSNumber KN5249500
TSCA No