missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Mercaptan 97.0+%, TCI America™

Catalog No. p-7118612
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068625ML 25 mL
B0686500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B068625ML Supplier TCI America Supplier No. B068625ML
Only null left

Chemical Identifiers

CAS 513-53-1
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00004865
InChI Key LOCHFZBWPCLPAN-UHFFFAOYSA-N
Synonym 2-butanethiol, sec-butyl mercaptan, 2-butyl mercaptan, sec-butanethiol, sec-butyl thiol, 2-mercaptobutane, 1-methyl-1-propanethiol, sec-butyl thioalcohol, secondary butylmercaptan, +/--1-methyl-1-propanethiol
PubChem CID 10560
IUPAC Name butane-2-thiol
SMILES CCC(C)S

Specifications

Melting Point -165°C
Color Yellow
Boiling Point 84°C
Quantity 25 mL
UN Number 2347
Formula Weight 90.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Mercaptan
EINECSNumber (2)-0464
TSCA Yes