accessibility menu, dialog, popup

UserName

CAS RN 513-53-1

H3CCH3HS

CAS RN 513-53-1

IUPAC Name: butane-2-thiol
Synonyms: 2-butanethiol
Molecular Weight (g/mol): 90.18
Molecular Formula: C4H10S
InChi Key: LOCHFZBWPCLPAN-UHFFFAOYNA-N
SMILES: CCC(C)S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results