missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Mercaptan 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | -165°C |
| Color | Yellow |
| Boiling Point | 84°C |
| Quantity | 500 mL |
| UN Number | 2347 |
| Formula Weight | 90.18 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | sec-Butyl Mercaptan |
Safety and Handling
| EINECSNumber | (2)-0464 |
| TSCA | Yes |