missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Ethyl Ether 97.0+%, TCI America™
$247.92 - $1487.52
Chemical Identifiers
| CAS | 2679-87-0 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00053773 |
| InChI Key | VSCUCHUDCLERMY-UHFFFAOYNA-N |
| PubChem CID | 17586 |
| IUPAC Name | 2-ethoxybutane |
| SMILES | CCOC(C)CC |
Chemical Identifiers
| 2679-87-0 | |
| 102.18 | |
| VSCUCHUDCLERMY-UHFFFAOYNA-N | |
| 2-ethoxybutane |
| C6H14O | |
| MFCD00053773 | |
| 17586 | |
| CCOC(C)CC |
Specifications
| 2679-87-0 | |
| 82°C | |
| MFCD00053773 | |
| 1179 | |
| CCOC(C)CC | |
| 102.18 | |
| 102.18 | |
| Liquid |
| Colorless | |
| C6H14O | |
| 25 g | |
| VSCUCHUDCLERMY-UHFFFAOYNA-N | |
| 2-ethoxybutane | |
| 17586 | |
| ≥97.0% (GC) | |
| sec-Butyl Ethyl Ether |
Safety and Handling
RTECSNumber : KN4730000
TSCA : Yes