accessibility menu, dialog, popup

UserName

CAS RN 2679-87-0

H3COCH3CH3

CAS RN 2679-87-0

IUPAC Name: 2-ethoxybutane
Synonyms: butane, 2-ethoxy-
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: VSCUCHUDCLERMY-UHFFFAOYNA-N
SMILES: CCOC(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results