missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Butyrate 98.0+%, TCI America™

Catalog No. p-7123017
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35595ML 5 mL
B355925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35595ML Supplier TCI America Supplier No. B35595ML
Only null left

Chemical Identifiers

CAS 819-97-6
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD15072150
InChI Key QJHDFBAAFGELLO-UHFFFAOYNA-N
Synonym Butyric Acid sec-Butyl Ester
PubChem CID 13174
ChEBI CHEBI:87318
IUPAC Name butan-2-yl butanoate
SMILES CCCC(=O)OC(C)CC

Specifications

Color Colorless
Boiling Point 152°C
Quantity 5 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Butyrate
EINECSNumber (2)-0779
TSCA No