accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 819-97-6

IUPAC Name: butan-2-yl butanoate
Synonyms: sec-butyl butanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: QJHDFBAAFGELLO-UHFFFAOYNA-N
SMILES: CCCC(=O)OC(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results